Learn More
Invitrogen™ mCherry Polyclonal Antibody

Description
Application notes: Indirect ELISA.
Specifications
Specifications
| Antigen | mCherry |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 50% glycerol, 0.2% BSA and 0.05% sodium azide; pH 7.4 |
| Gene Alias | mCherry |
| Host Species | Rabbit |
| Immunogen | Recombinant protein within mCherry 1-236 aa. |
| Purification Method | Antigen affinity chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.