Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ mCherry Polyclonal Antibody

Catalog No. PIPA5143556
Change view
Click to view available options
Quantity:
100 μL
500 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143556 500 μL
PIPA5143590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIPA5143556 Supplier Invitrogen™ Supplier No. PA5143556
Add to Cart
Add to Cart

Goat Polyclonal Antibody

mCherry Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen mCherry
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 5mM sodium azide
Gene Alias mCherry
Host Species Goat
Immunogen Full length recombinant mCherry protein.
Purification Method Antigen affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.