Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ mCherry Polyclonal Antibody
Goat Polyclonal Antibody
$527.50 - $1763.00
Specifications
Antigen | mCherry |
---|---|
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5143556
|
Invitrogen™
PA5143556 |
500 μL |
Each for $1,763.00
|
|
|||||
PIPA5143590
|
Invitrogen™
PA5143590 |
100 μL |
Each for $527.50
|
|
|||||
Description
mCherry Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.Specifications
mCherry | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Chemical | |
mCherry | |
Primary | |
Affinity chromatography |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Goat | |
RUO | |
PBS with 50% glycerol and 5mM sodium azide | |
Full length recombinant mCherry protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title