Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mCherry Rabbit anti-Tag, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$457.98
Specifications
Antigen | mCherry |
---|---|
Concentration | 0.41 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-5448
![]() ![]() |
Proteintech Group Inc
267651AP150UL |
150 μL |
Each of 1 for $457.98
|
|
|||||
Description
MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0.5 of 15 minutes, allowing it to be visualised soon after translation.Specifications
mCherry | |
Western Blot | |
Unconjugated | |
Rabbit | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
mCherry Fusion Protein Ag25320 | |
Primary | |
-20°C |
0.41 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
mCherry | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title