Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCHR Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987384
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7384 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7384 Supplier Bioss Supplier No. BS6626R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MCHR1 is a member of the G protein-coupled receptor family 1. It is an integral plasma membrane protein which binds melanin-concentrating hormone. The encoded protein can inhibit cAMP accumulation and stimulate intracellular calcium flux, and is probably involved in the neuronal regulation of food consumption. Although structurally similar to somatostatin receptors, this protein does not seem to bind somatostatin.

Specifications

Antigen MCHR
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Mchr1
Gene Accession No. Q99705
Gene Alias AW049955; G protein coupled receptor 24; G protein-coupled receptor 24; GPCR24; Gpr24; Gpr24-9 protein coupled receptor; G-protein coupled receptor 24; MCH 1 receptor; MCH receptor 1; MCH1; MCH1R; MCH-1R; MCHR; Mchr1; MCH-R1; MCHR-1; melanin concentrating hormone receptor 1; Melanin-concentrating hormone receptor 1; SLC1; SLC-1; Somatostatin receptor-like protein
Gene Symbols Mchr1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MCHR.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2847
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.