Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCHR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102101 Supplier Invitrogen™ Supplier No. PA5102101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MCHR2.

Melanin-concentrating hormone (MCH) is a 19 amino acid cyclic neuropeptide which is mainly expressed in the hypothalamus. MCH modulates feeding behavior, aggression, anxiety, arousal and reproductive function in mammals by controlling the release of luteinizing hormone (LH). The melanin-concentrating hormone receptor (MCHR, also designated SLC-1) is a glycosylated G-protein coupled receptor that migrates between 45-50 kDa on SDS-PAGE. MCHR mediates the effects of MCH through Gai and/or Gaq signaling and is expressed in several regions of the brain, including the cerebral cortex, amygdala, thalamus and hypothalamus. MCH and MCHR have also been implicated in stimulating leptin expression and secretion in adipocytes, which suggests that the melanin-concentrating hormone and its receptor may be potential targets for modulating obesity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCHR2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene MCHR2
Gene Accession No. Q969V1
Gene Alias G protein-coupled receptor 145; G protein-coupled receptor slt; GPR145; G-protein coupled receptor 145; GPRv17; MCH receptor 2; MCH2; Mch2 receptor; MCH2R; MCH-2R; MCHR2; MCH-R2; MCHR-2; melanin concentrating hormone receptor 2; melanin-concentrating hormone 2; melanin-concentrating hormone receptor 2; SLT; SLT receptor
Gene Symbols MCHR2
Host Species Rabbit
Immunogen A synthesized peptide derived from human MCHR2(Accession Q969V1), corresponding to amino acid residues S299-F340.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84539
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.