Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCHR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA131050
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA131050 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA131050 Supplier Invitrogen™ Supplier No. PA131050
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-31050 detects GPCR MCH2 from human samples.

Melanin-concentrating hormone (MCH) is a 19 amino acid cyclic neuropeptide which is mainly expressed in the hypothalamus. MCH modulates feeding behavior, aggression, anxiety, arousal and reproductive function in mammals by controlling the release of luteinizing hormone (LH). The melanin-concentrating hormone receptor (MCHR, also designated SLC-1) is a glycosylated G-protein coupled receptor that migrates between 45-50 kDa on SDS-PAGE. MCHR mediates the effects of MCH through Gai and/or Gaq signaling and is expressed in several regions of the brain, including the cerebral cortex, amygdala, thalamus and hypothalamus. MCH and MCHR have also been implicated in stimulating leptin expression and secretion in adipocytes, which suggests that the melanin-concentrating hormone and its receptor may be potential targets for modulating obesity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCHR2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene MCHR2
Gene Accession No. Q969V1
Gene Alias G protein-coupled receptor 145; G protein-coupled receptor slt; GPR145; G-protein coupled receptor 145; GPRv17; MCH receptor 2; MCH2; Mch2 receptor; MCH2R; MCH-2R; MCHR2; MCH-R2; MCHR-2; melanin concentrating hormone receptor 2; melanin-concentrating hormone 2; melanin-concentrating hormone receptor 2; SLT; SLT receptor
Gene Symbols MCHR2
Host Species Rabbit
Immunogen Synthetic 19 amino acid peptide from C-terminus of human MCHR2.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84539
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.