Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCK10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115105
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115105 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115105 Supplier Thermo Scientific Supplier No. PA5115105
Add to Cart
Add to Cart

MCK10 Polyclonal Antibody

MCK10 Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)

TRUSTED_SUSTAINABILITY

Specifications

Antigen MCK10
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene DDR1
Gene Accession No. Q03146, Q08345, Q63474
Gene Alias 6030432F18; AI323681; Cak; CD167; CD167 antigen-like family member A; CD167a; Cell adhesion kinase; DAAP-278B20.1; DDR; DDR1; DDR1d; discoidin domain receptor family, member 1; discoidin domain receptor tyrosine kinase 1; discoidin receptor tyrosine kinase; Drd1; Eddr1; Epithelial discoidin domain receptor 1; epithelial discoidin domain-containing receptor 1; HGK2; mammary carcinoma kinase 10; MCK10; MCK-10; Mpk6; NEP; neuroepithelial tyrosine kinase; neurotrophic tyrosine kinase receptor type 4; neurotrophic tyrosine kinase, receptor, type 4; NTRK4; protein-tyrosine kinase 3; Protein-tyrosine kinase 3A; Protein-tyrosine kinase MPK-6; protein-tyrosine kinase PTK-3; protein-tyrosine kinase RTK-6; PTK3; PTK3A; PTK3A protein tyrosine kinase 3A; PTK3D; RTK6; TRK E; TRKE; Tyrosine kinase DDR; tyrosine-protein kinase CAK
Gene Symbols DDR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human DDR1(Accession Q08345), corresponding to amino acid residues N771-T821.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12305, 25678, 780
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.