Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCK10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532718
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532718 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532718 Supplier Invitrogen™ Supplier No. PA532718
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon, Monkey, Marmoset, Dog, Bat, Bovine, Panda, Horse, Pig, Opossum (100%); Mouse, Rat, Hamster, Rabbit (94%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

DDR1 is a receptor tyrosine kinase activated by collagen and is reported to regulate adhesion, cell migration and other cellular processes. Receptor tyrosine kinases (RTKs) play a key role in the communication of cells with their microenvironment. These molecules are involved in the regulation of cell growth, differentiation and metabolism. The protein encoded by this gene is a RTK that is widely expressed in normal and transformed epithelial cells and is activated by various types of collagen. This protein belongs to a subfamily of tyrosine kinase receptors with a homology region to the Dictyostelium discoideum protein discoidin I in their extracellular domain. Its autophosphorylation is achieved by all collagens so far tested (type I to type VI). In situ studies and Northern-blot analysis showed that expression of this encoded protein is restricted to epithelial cells, particularly in the kidney, lung, gastrointestinal tract, and brain. In addition, this protein is significantly over-expressed in several human tumors from breast, ovarian, esophageal, and pediatric brain. This gene is located on chromosome 6p21.3 in proximity to several HLA class I genes. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCK10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene DDR1
Gene Accession No. Q08345
Gene Alias 6030432F18; AI323681; Cak; CD167; CD167 antigen-like family member A; CD167a; Cell adhesion kinase; DAAP-278B20.1; DDR; DDR1; DDR1d; discoidin domain receptor family, member 1; discoidin domain receptor tyrosine kinase 1; discoidin receptor tyrosine kinase; Drd1; Eddr1; Epithelial discoidin domain receptor 1; epithelial discoidin domain-containing receptor 1; HGK2; mammary carcinoma kinase 10; MCK10; MCK-10; Mpk6; NEP; neuroepithelial tyrosine kinase; neurotrophic tyrosine kinase receptor type 4; neurotrophic tyrosine kinase, receptor, type 4; NTRK4; protein-tyrosine kinase 3; Protein-tyrosine kinase 3A; Protein-tyrosine kinase MPK-6; protein-tyrosine kinase PTK-3; protein-tyrosine kinase RTK-6; PTK3; PTK3A; PTK3A protein tyrosine kinase 3A; PTK3D; RTK6; TRK E; TRKE; Tyrosine kinase DDR; tyrosine-protein kinase CAK
Gene Symbols DDR1
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human DDR1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100050861, 100144462, 100856512, 534092, 780
Target Species Human, Canine, Monkey, Bovine, Pig, Horse, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.