Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mcl-1 Antibody (OTI10F6), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272598MFV610
Description
Mcl-1 Monoclonal antibody specifically detects Mcl-1 in Human samples. It is validated for Flow CytometrySpecifications
Mcl-1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
OTI10F6 | |
Flow Cytometry | |
bcl2-L-3, BCL2L3MGC104264, Bcl-2-like protein 3, Bcl-2-related protein EAT/mcl1, EAT, induced myeloid leukemia cell differentiation protein Mcl-1, Mcl-1, mcl1/EAT, MCL1-ES, MCL1L, MCL1S, MGC1839, myeloid cell leukemia ES, myeloid cell leukemia sequence 1 (BCL2-related), TM | |
Full length human recombinant protein of human MCL1 (NP_068779) produced in HEK293T cell. | |
0.1 mL | |
Apoptosis, Cancer, Signal Transduction, Tumor Suppressors | |
4170 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction