Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCM-BP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | MCM-BP |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-5242
![]() |
Bethyl Laboratories, Inc
A303477AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt.
CJ119 is a carefully orchestrated process involving many proteins that assemble at origins of replication. Among these are the 6 proteins of the minichromosome maintece (MCM) complex (e.g., MCM2; MIM 116945), which form a hexamer. Each MCM subunit performs an essential function in initiation and elongation of DNA replication. MCMBP can replace MCM2 in the MCM complex, thus forming an alternative MCM hexamer.Specifications
MCM-BP | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
C10orf119, MCM-BP, MCM-binding protein, mini-chromosome maintece complex-binding protein, minichromosome maintece complex-binding protein | |
MCMBP | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q9BTE3 | |
79892 | |
Between 150 and 200 | |
Primary | |
4° C | |
MCMBP |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title