Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM-BP Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501565240
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-5240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-5240 Supplier Bethyl Laboratories, Inc Supplier No. A303477A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

CJ119 is a carefully orchestrated process involving many proteins that assemble at origins of replication. Among these are the 6 proteins of the minichromosome maintece (MCM) complex (e.g., MCM2; MIM 116945), which form a hexamer. Each MCM subunit performs an essential function in initiation and elongation of DNA replication. MCMBP can replace MCM2 in the MCM complex, thus forming an alternative MCM hexamer.

Specifications

Antigen MCM-BP
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene MCMBP
Gene Accession No. Q9BTE3
Gene Alias C10orf119, MCM-BP, MCM-binding protein, mini-chromosome maintece complex-binding protein, minichromosome maintece complex-binding protein
Gene Symbols MCMBP
Host Species Rabbit
Immunogen Between 150 and 200
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79892
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.