Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCM10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596321
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596321 Supplier Invitrogen™ Supplier No. PA596321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLEGAPATMT PKLGRGVLEG DDVLFYDESP PPRPKLSALA EAKKLAAITK LRAKGQVLTK TNPNSIKKKQ KDPQDILEVK ERVEKNTMFS SQAEDELEPA RKKRREQLAY LESEEFQKIL KAKSKHTGIL KEAEAEMQER YFEPLVKKEQ MEEKMRNIRE VKCRVVTCKT CAYTHFKLLE TCVSEQHEYH WHDGVKRFFK CPCGNRSISL DRLPNKHCSN CGLYKWERDG MLKEKTGPKI GGETLLPRGE EHAKFLNSLK; Positive Samples: Mouse heart; Cellular Location: Nucleus.

The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are involved in the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre-RC) and it may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein can interact with MCM2 and MCM6, as well as with the origin recognition protein ORC2. It is regulated by proteolysis and phosphorylation in a cell cycle-dependent manner. Studies of a similar protein in Xenopus suggest that the chromatin binding of this protein at the onset of DNA replication is after pre-RC assembly and before origin unwinding. Alternatively spliced transcript variants encoding distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCM10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MCM10
Gene Accession No. Q0VBD2, Q7L590
Gene Alias 2410041F14Rik; AU018508; C330019M07Rik; C79164; CNA43; DNA43; homolog of yeast MCM10; hsMCM10; Mcm10; MCM10 minichromosome maintenance deficient 10; MGC126776; minichromosome maintenance 10 replication initiation factor; minichromosome maintenance complex component 10; minichromosome maintenance deficient 10; minichromosome maintenance deficient 10 (S. cerevisiae); PRO2249; Protein MCM10 homolog
Gene Symbols MCM10
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55388, 70024
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.