Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MCM10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541323
Description
Peptide sequence: PKLSALAEAK KLAAITKLRA KGQVLTKTNP NSIKKKQKDP QDILEVKERV Sequence homology: Cow: 77%; Dog: 86%; Horse: 93%; Human: 100%; Pig: 77%; Rabbit: 86%.
The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are involved in the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre-RC) and it may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein can interact with MCM2 and MCM6, as well as with the origin recognition protein ORC2. It is regulated by proteolysis and phosphorylation in a cell cycle-dependent manner. Studies of a similar protein in Xenopus suggest that the chromatin binding of this protein at the onset of DNA replication is after pre-RC assembly and before origin unwinding. Alternatively spliced transcript variants encoding distinct isoforms have been identified.
Specifications
| MCM10 | |
| Polyclonal | |
| Unconjugated | |
| MCM10 | |
| 2410041F14Rik; AU018508; C330019M07Rik; C79164; CNA43; DNA43; homolog of yeast MCM10; hsMCM10; Mcm10; MCM10 minichromosome maintenance deficient 10; MGC126776; minichromosome maintenance 10 replication initiation factor; minichromosome maintenance complex component 10; minichromosome maintenance deficient 10; minichromosome maintenance deficient 10 (S. cerevisiae); PRO2249; Protein MCM10 homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55388 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q7L590 | |
| MCM10 | |
| Synthetic peptide directed towards the middle region of human MCM10. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction