Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCM2 Mouse anti-Human, Clone: 1F4D10, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | MCM2 |
---|---|
Clone | 1F4D10 |
Concentration | 0.8 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6637
![]() ![]() |
Proteintech Group Inc
662041IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
MCM2 (Mini chromosome maintece protein 2) is involved in regulating DNA replication. MCM2 (also called CDCL1, mitotin and BM28), is a human nuclear protein that is crucial in the cell cycle, being involved in the onset of DNA replication and cell division. It is similar to members of the family of early S-phase proteins. MCM2 has been mapped to 3q21. From its localization, CDCL1 became a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML).Specifications
MCM2 | |
0.8 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BM28, CCNL1, cdc19, CDCL1, D3S3194, KIAA0030, MCM2, MITOTIN, Nuclear protein BM28 | |
MCM2 | |
IgG1 | |
Protein G | |
Antibody |
1F4D10 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P49736 | |
4171 | |
MCM2 Fusion Protein Ag0798 | |
Primary | |
-20°C | |
MCM2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title