Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM2 Rabbit anti-Human, Unconjugated, Polyclonal, Active Motif™
SDP

Catalog No. 501993648
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3648 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-3648 Supplier Active Motif Supplier No. 61717
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MCM2 (Mini chromosome maintenance protein 2) is involved in regulating DNA replication. MCM2 (also called CDCL1, mitotin and BM28), is a human nuclear protein that is crucial in the cell cycle, being involved in the onset of DNA replication and cell division. It is similar to members of the family of early S-phase proteins. MCM2 has been mapped to 3q21. From its localization, CDCL1 became a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML).

Specifications

Antigen MCM2
Applications Western Blot
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.5
Gene MCM2
Gene Accession No. P49736
Gene Alias AA959861; AW476101; BM28; CCNL1; cdc19; Cdcl1; cell devision cycle-like 1; cyclin-like 1; D3S3194; DFNA70; DNA replication licensing factor MCM2; KIAA0030; Mcm2; Mcmd2; MGC10606; mini chromosome maintenance deficient 2; minichromosome maintenance complex component 2; minichromosome maintenance deficient 2 (mitotin); minichromosome maintenance deficient 2 mitotin; minichromosome maintenance deficient 2 mitotin (S. cerevisiae); Minichromosome maintenance protein 2 homolog; MITOTIN; mKIAA0030; Nuclear protein BM28
Gene Symbols MCM2
Host Species Rabbit
Immunogen This antibody was raised against a full-length recombinant protein corresponding to human MCM2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4171
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.