Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCM3AP Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171883F
Description
MCM3AP Polyclonal antibody specifically detects MCM3AP in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
MCM3AP | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
80 kDa MCM3-associated protein, FLJ44336, GANPFLJ45306, human mRNA for MCM3 import factor10Protein GANP, KIAA0572MAP80germinal center-associated nuclear protein, Map80, MCM3 import protein, MCM3 minichromosome maintenance deficient 3 (S. cerevisiae) associated protein, MCM3 minichromosome maintenance deficient 3 associated protein, minichromosome maintenance complex component 3 associated protein, minichromosome maintenance deficient (S. cerevisiae) 3-associated protein, minichromosome maintenance deficient 3-associated protein | |
The immunogen for this product maps to a region between residue 1930 and 1980 of human Germinal center-Associated Nuclear Protein using the numbering given in entry NP_003897.2 (GeneID 8888). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
FITC | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
8888 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction