Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCM4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568750 Supplier Invitrogen™ Supplier No. PA568750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.

The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 6 and 7 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. The phosphorylation of this protein by CDC2 kinase reduces the DNA helicase activity and chromatin binding of the MCM complex. This gene is mapped to a region on the chromosome 8 head-to-head next to the PRKDC/DNA-PK, a DNA-activated protein kinase involved in the repair of DNA double-strand breaks. Alternatively spliced transcript variants encoding the same protein have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCM4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Mcm4
Gene Accession No. 0
Gene Alias 19G; AI325074; AU045576; cb1025; Cdc21; CDC21 homolog; CDC54; DNA replication licensing factor MCM4; fc12c09; fj85g09; hCdc21; hm:zeh1616; homolog of S. pombe cell devision cycle 21; mcdc21; MCM4; MCM4 minichromosome maintenance deficient 4; MCM4 minichromosome maintenance deficient 4, mitotin; mcm4 protein; Mcmd4; mini chromosome maintenance deficient 4 homolog; minichromosome maintenance complex component 4; minichromosome maintenance deficient 4; minichromosome maintenance deficient 4 homolog; minichromosome maintenance deficient 4 homolog (S. cerevisiae); mKIAA4003; NKCD; NKGCD; P1-CDC21; Unknown (protein for MGC:140484); wu:fc12c09; wu:fj85g09; zeh1616
Gene Symbols Mcm4
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human MCM4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29728
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.