Learn More
MCM4 Polyclonal Antibody, Bethyl Laboratories
Goat Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | MCM4 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1374
![]() |
Bethyl Laboratories, Inc
IHC00038T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1373
![]() |
Bethyl Laboratories, Inc
IHC00038 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rabbit, Guinea pig_10141, Orangutan, Monkey, Gorilla, Chimpanzee, Northern white-cheeked g
The protein encoded by this gene is one of the highly conserved mini-chromosome maintece proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 6 and 7 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. The phosphorylation of this protein by CDC2 kinase reduces the DNA helicase activity and chromatin binding of the MCM complex. This gene is mapped to a region on the chromosome 8 head-to-head next to the PRKDC/DNA-PK, a DNA-activated protein kinase involved in the repair of DNA double-strand breaks. Alternatively spliced transcript variants encoding the same protein have been reported.Specifications
MCM4 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
CDC21, CDC54, P1-CDC21, hCdc21, CDC21 homolog, DNA replication licensing factor MCM4, MCM4 minichromosome maintece deficient 4, homolog of S. pombe cell devision cycle 21, minichromosome maintece deficient 4 | |
MCM4 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Goat | |
Human | |
P33991 | |
4173 | |
Between 325 and 375 | |
Primary | |
4° C | |
MCM4 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.