Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCM5 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10079778AF647
Description
MCM5 Polyclonal antibody specifically detects MCM5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
MCM5 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
CDC46 homolog, CDC46DNA replication licensing factor MCM5, EC 3.6.4.12, MCM5 minichromosome maintenance deficient 5, cell division cycle 46, MCM5 minichromosome maintenance deficient 5, cell division cycle 46 (S.cerevisiae), MGC5315, minichromosome maintenance complex component 5, minichromosome maintenance deficient (S. cerevisiae) 5 (cell division cycle 46), minichromosome maintenance deficient 5 (cell division cycle 46), P1-CDC46 | |
The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Minichromosome Maintenance 5 using the numbering given in entry NP_006730.2 (GeneID 4174). | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
4174 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction