Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCM5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120220
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120220 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120220 Supplier Invitrogen™ Supplier No. PA5120220
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Raji, HL-60, HeLa, SW620, Mouse kidney, Rat kidney. The target is usually found in the following locations: Nucleus. Immunogen sequence: MSGFDDPGIF YSDSFGGDAQ ADEGQARKSQ LQRRFKEFLR QYRVGTDRTG FTFKYRDELK RHYNLGEYWI EVEMEDLASF DEDLADYLYK QPAEHLQLLE EAAKEVADEV TRPRPSGEEV LQDIQVMLKS DASPSSIRSL KSDMMSHLVK IPGIIIAASA VRAKATRISI QCRSCRNTLT NIAMRPGLEG YALPRKCNTD QAGRPKCPLD PYFIMPDKCK CVDFQTLKLQ.

The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCM5
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MCM5
Gene Accession No. P33992, P49718
Gene Alias AA617332; AI324988; AL033333; Cdc46; CDC46 homolog; DNA replication licensing factor MCM5; MCM5; MCM5 minichromosome maintenance deficient 5, cell division cycle 46; Mcmd5; MGC5315; minichromosome maintenance complex component 5; minichromosome maintenance deficient 5 (cell division cycle 46); minichromosome maintenance deficient 5, cell division cycle 46; minichromosome maintenance deficient 5, cell division cycle 46 (S. cerevisiae); P1-CDC46
Gene Symbols MCM5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human MCM5 (NP_006730.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17218, 291885, 4174
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.