Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCM6 Antibody, Alexa Fluor™ 594, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB100251AF594
Description
MCM6 Polyclonal antibody specifically detects MCM6 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
MCM6 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
DNA replication licensing factor MCM6, EC 3.6.4.12, MCG40308, MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe), MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe) (S.cerevisiae), minichromosome maintenance complex component 6, minichromosome maintenance deficient (mis5, S. pombe) 6, minichromosome maintenance deficient 6 homolog, minichromosome maintenance deficient 6 homolog (S. cerevisiae), Mis5, MIS5 homolog, p105MCM | |
The immunogen recognized by this antibody maps to a region between residue 775 and the C-terminus (residue 821) of human Minichromosome Maintenance 6 using the numbering given in entry NP_005906.2 (GeneID 4175). | |
0.1 mL | |
Cell Cycle and Replication, DNA Repair | |
4175 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Goat | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction