Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCM6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578056
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578056 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578056 Supplier Invitrogen™ Supplier No. PA578056
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Mouse (97%), Rat (97%), Chicken (90%), Rhesus Monkey (100%), Bovine (97%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

MCM6 may be involved in the control of a single round of DNA replication during S phase. It binds to chromatin during G1 and detaches from it during S phase as if it licenses the chromatin to replicate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCM6
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.14 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene Mcm6
Gene Accession No. Q14566
Gene Alias ASP-l1; D1Wsu22e; DNA replication licensing factor MCM6; intestinal DNA replication protein; MCG40308; Mcm6; MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe); Mcmd6; mini chromosome maintenance deficient 6; minichromosome maintenance complex component 6; minichromosome maintenance deficient (mis5, S. pombe) 6; minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe); minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe) (S. cerevisiae); Mis5; Mis5 homolog; p105MCM
Gene Symbols Mcm6
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human MCM6. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4175
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.