Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCM6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596010 Supplier Invitrogen™ Supplier No. PA596010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SIDRVYSLDD IRRYLLFARQ FKPKISKESE DFIVEQYKHL RQRDGSGVTK SSWRITVRQL ESMIRLSEAM ARMHCCDEVQ PKHVKEAFRL LNKSIIRVET PDVNLDQEEE IQMEVDEGAG GINGHADSPA PVNGINGYNE DINQESAPKA SLRLGFSEYC RISNLIVLHL RKVEEEEDES ALKRSELVNW YLKEIESEID SEEELINKKR IIEKVIHRLT HYDHVLIELT QAGLKGSTEG SESYEEDPYL VVNPNYLLED; Positive Samples: U-251MG, HeLa, MCF7, HL-60, NIH/3T3, Mouse spleen; Cellular Location: Nucleus.

MCM6 may be involved in the control of a single round of DNA replication during S phase. It binds to chromatin during G1 and detaches from it during S phase as if it licenses the chromatin to replicate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCM6
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Mcm6
Gene Accession No. P97311, Q14566, Q62724
Gene Alias ASP-l1; D1Wsu22e; DNA replication licensing factor MCM6; intestinal DNA replication protein; MCG40308; Mcm6; MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe); Mcmd6; mini chromosome maintenance deficient 6; minichromosome maintenance complex component 6; minichromosome maintenance deficient (mis5, S. pombe) 6; minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe); minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe) (S. cerevisiae); Mis5; Mis5 homolog; p105MCM
Gene Symbols Mcm6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 562-821 of human MCM6 (NP_0059062).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17219, 29685, 4175
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.