Learn More
MCM6 Goat anti-Human, Polyclonal, Bethyl Laboratories
Goat Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300127A
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Bovine
The protein encoded by this gene is one of the highly conserved mini-chromosome maintece proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 4 and 7 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. The phosphorylation of the complex by CDC2 kinase reduces the helicase activity, suggesting a role in the regulation of DNA replication.Specifications
| MCM6 | |
| Polyclonal | |
| Unconjugated | |
| MCM6 | |
| MCG40308, Mis5, P105MCM | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 4175 | |
| 4° C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q14566 | |
| MCM6 | |
| Between 775 and C-term | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.