Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM6 Rabbit anti-Human, Polyclonal, Abnova™

Catalog No. 89033209 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-033-209 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-033-209 Supplier Abnova Corporation Supplier No. PAB29921
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rabbit polyclonal antibody raised against partial synthetic protein of human MCM6.

  • High specificity
  • High reactivity

Specifications

Antigen MCM6
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial synthetic protein of human MCM6.
Dilution Immunohistochemistry (1:250) Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In 1X PBS , pH 7.4 (2% sucrose, 0.09% sodium azide).
Gene Accession No. Q14566
Gene Alias MCG40308/Mis5/P105MCM
Gene Symbols MCM6
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 771-820 of human MCM6.
Purification Method Affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4175
Target Species Human
Content And Storage Store at 4°C for up to one week. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.