Learn More
MCM6 Recombinant Rabbit Monoclonal Antibody (JE65-68), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Supplier: Thermo Scientific MA550666

Description
Positive control: Hela cell lysate, Jurkat cell lysate, MCF-7 cell lysate, RAW264.7 cell lysate, SiHa, rat skin tissue, human lymph nodes tissue, human testis tissue, mouse stomach tissue. Predicted band size: 93 kDa Subcellular Location: Nucleus, Chromosome.
MCM6 may be involved in the control of a single round of DNA replication during S phase. It binds to chromatin during G1 and detaches from it during S phase as if it licenses the chromatin to replicate.
Specifications
MCM6 | |
Recombinant Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P97311, Q14566, Q62724 | |
Mcm6 | |
Recombinant protein within human MCM6 aa 572-821/821. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
JE65-68 | |
Unconjugated | |
Mcm6 | |
ASP-l1, D1Wsu22e, DNA replication licensing factor MCM6, intestinal DNA replication protein, MCG40308, Mcm6, MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe), Mcmd6, mini chromosome maintenance deficient 6, minichromosome maintenance complex component 6, minichromosome maintenance deficient (mis5, S. pombe) 6, minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe), minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe) (S. cerevisiae), Mis5, Mis5 homolog, p105MCM | |
Rabbit | |
Protein A | |
RUO | |
17219, 29685, 4175 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.