Learn More
MCM7 Antibody (2068B), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB9217MFV500
Description
MCM7 Monoclonal antibody specifically detects MCM7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, ImmunocytochemistrySpecifications
| MCM7 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 4176 | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
| 2068B | |
| Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47 | |
| E. coli-derived recombinant human MCM7, Gly188-Ala328, Accession # P33993 | |
| RUO | |
| Primary | |
| Human | |
| IgG |