Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCM7 Antibody (MCM7/2756R), DyLight 680, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP308404FR
Description
MCM7 Monoclonal antibody specifically detects MCM7 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
MCM7 | |
Monoclonal | |
DyLight 680 | |
CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47 | |
Recombinant fragment of human MCM7 (aa195-319) (Uniprot: P33993) | |
100 μg | |
Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
4176 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
MCM7/2756R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction