Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM7 Monoclonal Antibody (47DC141 (DCS-141)), Invitrogen™

Catalog No. PIMA514291
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514291 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514291 Supplier Thermo Scientific Supplier No. MA514291

Mouse Monoclonal Antibody

MA5-14291 targets CDC47 in IF, IHC (P), IP, and WB applications and shows reactivity with Canine, Human, mouse, Rat, and Xenopus laevis samples. The MA5-14291 immunogen is recombinant hCDC47 protein.

MCM7 encodes a protein that is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 4 and 6 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme.The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 4 and 6 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. Cyclin D1-dependent kinase, CDK4, is found to associate with this protein, and may regulate the binding of this protein with the tumorsuppressor protein RB1/RB. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCM7
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 47DC141 (DCS-141)
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene Mcm7
Gene Accession No. P33993, Q61881, Q91876
Gene Alias AI747533; CDC47; CDC47 homolog; CDC47 homolog A; CDC47p; DNA replication licensing factor MCM7; DNA replication licensing factor mcm7-A; DNA replication licensing factor MCM7-like protein; homolog of S. cerevisiae Cdc47; mCDC47; MCM2; MCM7; MCM7 minichromosome maintenance deficient 7; mcm7.S; mcm7-a; mcm7-b; Mcmd7; mini chromosome maintenance deficient 7; minichromosome maintenance complex component 7; minichromosome maintenance complex component 7 S homeolog; minichromosome maintenance deficient 7; minichromosome maintenance deficient 7 (S. cerevisiae); minichromosome maintenance protein 7; minichromosome maintenance protein 7-A; P1 P85MCM; P1.1-MCM3; P1CDC47; P85MCM; p90; PNAS146; PPP1R104; protein phosphatase 1, regulatory subunit 104; Unknown (protein for MGC:128845); XELAEV_18000877mg; XMCM7; xMCM7-A
Gene Symbols Mcm7, mcm7.S
Host Species Mouse
Immunogen Recombinant hCDC47 protein
Purification Method Protein G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17220, 288532, 397852, 4176, 479737
Target Species Canine, Human, Mouse, Rat, X. laevis
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.