Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCM7 Mouse anti-Human, Clone: MCM7/1469, Abnova™

Catalog No. 89034675 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-034-675 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-034-675 Supplier Abnova Corporation Supplier No. MAB14592
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human MCM7.

  • High specificity
  • High reactivity

Specifications

Antigen MCM7
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone MCM7/1469
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human MCM7.
Dilution Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. P33993
Gene Alias CDABP0042/CDC47/MCM2/P1.1-MCM3/P1CDC47/P85MCM/PNAS-146
Gene Symbols MCM7
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 195-319 of human MCM7.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4176
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2b κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.