Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCM7 Mouse anti-Human, DyLight 488, Clone: 118H7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP260247G
This item is not returnable.
View return policy
Description
MCM7 Monoclonal antibody specifically detects MCM7 in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
MCM7 | |
Monoclonal | |
DyLight 488 | |
CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47 | |
Recombinant human MCM7 (1-414aa) purified from E. coli. | |
0.1 mL | |
Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
4176 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunofluorescence | |
118H7 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction