Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCM7 Mouse anti-Human, HRP, Clone: MCM7/1467, Novus Biologicals™

Mouse Monoclonal Antibody
$646.00
Specifications
Antigen | MCM7 |
---|---|
Clone | MCM7/1467 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | HRP |
Description
MCM7 Monoclonal specifically detects MCM7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MCM7 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
Mouse | |
Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47 | |
Recombinant human MCM7 protein fragment (aa195-319) (exact sequence is proprietary) | |
Primary | |
Store at 4°C in the dark. |
MCM7/1467 | |
Monoclonal | |
Purified | |
RUO | |
PBS | |
4176 | |
IgG2a κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title