Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCM9 Monoclonal Antibody (5H8E10)
GREENER_CHOICE

Catalog No. FEMA550281
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550281 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550281 Supplier Invitrogen™ Supplier No. MA550281
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MCM9 Monoclonal Antibody for Western Blot, IHC (P), ELISA

Component of the MCM8-MCM9 complex, a complex involved in homologous recombination repair following DNA interstrand cross-links and plays a key role during gametogenesis. The MCM8- MCM9 complex probably acts as a hexameric helicase downstream of the Fanconi anemia proteins BRCA2 and RAD51 and is required to process aberrant forks into homologous recombination substrates and to orchestrate homologous recombination with resection, fork stabilization and fork restart.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCM9
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 5H8E10
Concentration 1.227 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene MCM9
Gene Accession No. Q9NXL9
Gene Alias 9030408O17Rik; C6orf61; dJ329L24.1; dJ329L24.3; DNA helicase MCM9; DNA replication licensing factor MCM9; Gm235; hMCM9; MCM9; Mcmdc1; Minichromosome maintenance 9; minichromosome maintenance 9 homologous recombination repair factor; minichromosome maintenance complex component 9; mini-chromosome maintenance deficient 9; minichromosome maintenance deficient domain containing 1; mini-chromosome maintenance deficient domain-containing protein 1; ODG4; RGD1560557
Gene Symbols MCM9
Host Species Mouse
Immunogen Recombinant Human DNA helicase MCM9 protein (1-391AA).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254394
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.