Learn More
Invitrogen™ MCM9 Polyclonal Antibody

Description
MCM9 Polyclonal Antibody for ELISA, ICC/IF, Western Blot
Specifications
Specifications
| Antigen | MCM9 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | MCM9 |
| Gene Accession No. | Q2KHI9, Q9NXL9 |
| Gene Alias | 9030408O17Rik; C6orf61; dJ329L24.1; dJ329L24.3; DNA helicase MCM9; DNA replication licensing factor MCM9; Gm235; hMCM9; MCM9; Mcmdc1; Minichromosome maintenance 9; minichromosome maintenance 9 homologous recombination repair factor; minichromosome maintenance complex component 9; mini-chromosome maintenance deficient 9; minichromosome maintenance deficient domain containing 1; mini-chromosome maintenance deficient domain-containing protein 1; ODG4; RGD1560557 |
| Gene Symbols | MCM9 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.