Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MCM9, Rabbit, Unlabeled, Polyclonal,

Catalog No. ABE2603 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABE2603 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABE2603 Supplier MilliporeSigma™ Supplier No. ABE2603
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNA helicase MCM9 (EC 3.6.4.12; UniProt Q9NXL9; also known as hMCM9, Mini-chromosome maintenance deficient domain-containing protein 1) is encoded by the MCM9 (also known as C6orf61, MCMDC1, ODG4) gene (Gene ID 254394) in human. Mammalian minichromosome maintenance (MCM) protein homologues MCM8 and MCM9 play important roles in DNA replication elongation and prereplication complex (pre-RC) formation. MCM9 is found physically interacted with MCM8 and the interaction is essential for MCM9 cellular stability. MCM8-MCM9 complex is required for homologous recombination- (HR-) mediated repair of double-strand breaks (DSBs). In addition, the ATPase motifs of MCM8 and MCM9 (a.a. 523-530) are needed for MRN (MRE11-RAD50-NBS1) complex recruitment to DSBs sites. Cancer cells with MCM9 homozygous deletion display enhanced reduction in cell survival following exposure to interstrand-crosslinking (ICL) agents, such as cisplatin, due to impaired HR efficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCM9
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Rabbit polyclonal antibody serum with 0.05% Sodium Azide.
Gene MCM9, C6orf61, MCMDC1, ODG4
Gene Accession No. Q9NXL9
Host Species Rabbit
Immunogen Recombinant human MCM9 C-terminal fragment.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_060166
Test Specificity This rabbit polyclonal antiserum detected DNA damage-induced MCM9 nuclear foci formation in U2OS and wild-type, but not Mcm9-null MEFs (Park, J., et al. (2013). Mol. Cell. Biol. 33(8):1632-1644). The C-terminal end epitope region is present in all four human MCM9 spliced isoforms and the first two mouse MCM9 isoforms reported by UniProt (Q9NXL9 and Q2KHI9).
Target Species Human
Content And Storage Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Serum
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.