Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MCMBP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549723
Description
The antibody detects endogenous level of total MCMBP protein.
MEK1 (Mitogen activated protein kinase kinase 1) catalyzes the concomitant phosphorylation of a threonine and a tyrosine residue in a Thr-Glu-Tyr sequence located in MAP kinases. MEK1 activates ERK1 and ERK2 MAP kinases. Mitogen activated protein kinase kinase 2 (MEK2 or MAPKK2) is a member of a family of tyrosine/threonine protein kinases that activate the ERK1 and 2 and MAPK enzymes by phosphorylating both residues within the threonine/glutamate/tyrosine (TEY) motif in the activation loop. MEK1 and 2 are also activated by dual phosphorylation, which occurs on serine 218 and 222, in the activation loop of the MEK. Threonine 292 of MEK1 is phosphorylated by ERK 2, which serves as a negative feedback loop by suppressing activation of MEK1.
Specifications
MCMBP | |
Polyclonal | |
Unconjugated | |
MCMBP | |
1110007A13Rik; AU017382; BC025641; C10orf119; cj119; MCM (minichromosome maintenance deficient) binding protein; MCM-binding protein; Mcmbp; MCM-BP; minichromosome maintenance complex binding protein; Mini-chromosome maintenance complex-binding protein; minichromosome maintenance complex-binding protein; RGD1306730 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
309009, 79892 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
B1H268, Q9BTE3 | |
MCMBP | |
Synthetic peptide peptide corresponding to a region derived from 176-190 amino acids of human minichromosome maintenance complex binding protein. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction