Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Contactin-6 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF5890SP
This item is not returnable.
View return policy
Description
Contactin-6 Polyclonal specifically detects Contactin-6 in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
Contactin-6 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CNTN6, contactin 6, contactin-6, Contactin6, MGC133256, NB3 | |
Sheep | |
Affinity Purified | |
RUO | |
27255 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Q9JMB8 | |
CNTN6 | |
Chinese hamster ovary cell line CHO-derived recombinant mouse Contactin-6 Glu20-Ser998 Accession # Q9JMB8 | |
25 μg | |
Primary | |
Detects mouse Contactin-6 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human (rh) Contactin-6 is observed, and less than 5% cross-reactivity with recombinant mouse (rm) Contactin-3, rmContactin-4, rmContactin-5, and rhTAG-1 is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction