Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCP-1 Monoclonal Antibody (1A7B8)
GREENER_CHOICE

Catalog No. PIMA514767
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514767 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514767 Supplier Invitrogen™ Supplier No. MA514767
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-14767 targets MCP-1 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-14767 immunogen is purified recombinant fragment of human MCP-1 expressed in E. Coli.

This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils. It has been implicated in the pathogenesis of diseases characterized by monocytic infiltrates, like psoriasis, rheumatoid arthritis and atherosclerosis. It binds to chemokine receptors CCR2 and CCR4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCP-1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1A7B8
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 0.03% sodium azide
Gene Ccl2
Gene Accession No. P13500
Gene Alias Acidic seminal fluid protein; AI323594; C-C motif chemokine 2; C-C motif chemokine ligand 2; Ccl2; chemokine (C-C motif) ligand 2; GDCF-2; HC11; H-MCP-1; HSMCR30; immediate-early serum-responsive JE protein; immediate-early serum-responsive protein JE; Je; MCAF; Mcp1; MCP-1; MCP1A; MCP-1A; MGC9434; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; monocyte chemotactic and activating factor; Monocyte chemotactic protein 1; monocyte chemotactic protein 1A; Monocyte secretory protein JE; monocyte-chemoattractant protein-1 precursor; platelet-derived growth factor-inducible protein JE; RP23-350G1.3; SCYA2; Sigje; small inducible cytokine A2; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; small inducible gene JE; small-inducible cytokine A2; SMC-CF
Gene Symbols Ccl2
Host Species Mouse
Immunogen Purified recombinant fragment of human MCP-1 expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6347
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.