Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCP-2 Monoclonal Antibody (35509.11), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA124663
Description
MA1-24663 detects MCP2 from human samples. MA1-24663 has been successfully used in Western blot procedures. The MA1-24663 immunogen is recombit human monocyte chemotactic protein 2 (MCP2), expressed in E. coli.
This gene is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes, lymphocytes, basophils and eosinophils. By recruiting leukocytes to sites of inflammation this cytokine may contribute to tumor-associated leukocyte infiltration and to the antiviral state against HIV infection.
Specifications
MCP-2 | |
Monoclonal | |
0.5 mg/mL | |
CCL8 | |
HC14, MCP-2, MCP2, SCYA10, SCYA8 | |
Mouse | |
Protein A | |
RUO | |
6355 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Neutralization, Western Blot | |
35509.11 | |
Unconjugated | |
P80075 | |
CCL8 | |
Recombit human monocyte chemotactic protein 2 (MCP2), expressed in E. coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction