Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MCP-4 Monoclonal Antibody (3G4)
GREENER_CHOICE

Catalog No. PIM809
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIM809 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIM809 Supplier Invitrogen™ Supplier No. M809
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

There is one major form (long chain) and two minor forms (short chain and medium chain) of MCP-4 produced by differential signal peptide cleavage. Thus, the resulting MW of secreted MCP-4 ranges between ~8-9kD. Western blot analysis of recombinant MCP-4/CCL13 protein detects an ~17kD band, likely corresponding to a dimer of MCP-4. The M809 MCP-4 antibody (clone 3G4) has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody M810 (biotinylated conjugate of clone 8C12). The M809 MCP-4 antibody (biotinylated conjugate of clone 3G4) has also been used successfully in a sandwich ELISA as a detection antibody when paired with M810 (clone 8C12) as the coating antibody.

Monocyte Chemotactic Protein 4 (MCP-4), also called CCL13, is induced by inflammatory proteins such as IL-1 and TNFalpha. MCP-4 is a ligand for three different G protein coupled receptors, CCR2, CCR3 and CCR5. MCP-4 activates signaling in monocytes, T lymphocytes, eosinophils and basophils and this signaling is associated with the allergic response.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCP-4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3G4
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene CCL13
Gene Accession No. Q99616
Gene Alias C-C motif chemokine 13; C-C motif chemokine 13, long chain; C-C motif chemokine 13, medium chain; C-C motif chemokine 13, short chain; C-C motif chemokine ligand 13; CCL 13; CCL13; chemokine (C-C motif) ligand 13; CKb10; CK-beta-10; H-MCP-4; MCP4; MCP-4; Monocyte chemoattractant protein 4; monocyte chemotactic protein 4; NCC1; NCC-1; new CC chemokine 1; SCYA13; SCYL1; small inducible cytokine subfamily A (Cys-Cys), member 13; small-inducible cytokine A13
Gene Symbols CCL13
Host Species Mouse
Immunogen Recombinant CCL13 protein.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6357
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.