Learn More
Invitrogen™ MCP-4 Monoclonal Antibody (8C12)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ M810
Description
The M810 MCP-4 antibody (clone 8C12) has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody M809 (biotinylated conjugate of clone 3G4). The M810 MCP-4 antibody (biotinylated conjugate of clone 8C12) has also been used successfully in a sandwich ELISA as a detection antibody when paired with M809 (clone 3G4) as the coating antibody.
Monocyte Chemotactic Protein 4 (MCP-4), also called CCL13, is induced by inflammatory proteins such as IL-1 and TNFalpha. MCP-4 is a ligand for three different G protein coupled receptors, CCR2, CCR3 and CCR5. MCP-4 activates signaling in monocytes, T lymphocytes, eosinophils and basophils and this signaling is associated with the allergic response.
Specifications
MCP-4 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q99616 | |
CCL13 | |
Recombinant CCL13 protein. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
8C12 | |
Unconjugated | |
CCL13 | |
C-C motif chemokine 13; C-C motif chemokine 13, long chain; C-C motif chemokine 13, medium chain; C-C motif chemokine 13, short chain; C-C motif chemokine ligand 13; CCL 13; CCL13; chemokine (C-C motif) ligand 13; CKb10; CK-beta-10; H-MCP-4; MCP4; MCP-4; Monocyte chemoattractant protein 4; monocyte chemotactic protein 4; NCC1; NCC-1; new CC chemokine 1; SCYA13; SCYL1; small inducible cytokine subfamily A (Cys-Cys), member 13; small-inducible cytokine A13 | |
Mouse | |
Protein A | |
RUO | |
6357 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.