Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCPH1 Antibody (OTI2A3), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272600APC
Description
MCPH1 Monoclonal antibody specifically detects MCPH1 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
MCPH1 | |
Monoclonal | |
APC | |
PBS | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
OTI2A3 | |
Western Blot, Immunohistochemistry | |
BRCT-repeat inhibitor of TERT expression 1, BRIT1, FLJ12847, MCT, microcephalin, microcephalin 1, microcephaly, primary autosomal recessive 1 | |
Human recombinant protein fragment corresponding to amino acids 1-264 of human MCPH1 (NP_078872) produced in E.coli. | |
0.1 mL | |
DNA Repair | |
79648 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction