Learn More
MCPH1 Mouse anti-Human, Clone: OTI4A12, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804946

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This protein is implicated in chromosome condensation and DNA damage induced cellular responses, and may play a role in neurogenesis and regulation of the size of the cerebral cortex.Specifications
MCPH1 | |
Monoclonal | |
Unconjugated | |
MCPH1 | |
BRIT1, MCT | |
Mouse | |
Affinity Chromatography | |
RUO | |
79648 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI4A12 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8NEM0 | |
MCPH1 | |
Human recombit protein fragment corresponding to amino acids 1-264 of human MCPH1 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.