Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCPIP1/ZC3H12A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | MCPIP1/ZC3H12A |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
MCPIP1/ZC3H12A Polyclonal specifically detects MCPIP1/ZC3H12A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MCPIP1/ZC3H12A | |
Polyclonal | |
Rabbit | |
Cancer, Cell Biology, Editing and Processing Endonucleases, Immunology, Inflammation | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Reg1, Regnase-1, zinc finger CCCH-type containing 12A | |
ZC3H12A | |
IgG | |
Affinity Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Q5D1E8 | |
80149 | |
This antibody was developed against a recombinant protein corresponding to amino acids: CTYGIKCRFFHPERPSCPQRSVADELRANALLSPPRAPSKDKNGRRPSPSSQSSSLLTESEQCSLDGKKLGAQASPGSRQEGLTQTYAPSGRSLAP | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title