Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MCPT4 Polyclonal Antibody
MCPT4 Polyclonal Antibody
Supplier: Thermo Scientific PA5142454
This item is not returnable.
View return policy
Description
MCPT4 Polyclonal Antibody for IHC (P)

Specifications
MCPT4 | |
Polyclonal | |
Unconjugated | |
Mcpt4 | |
Mcpt4 | |
Peptide with sequence C-KLQKKAKETPSVN (aa 114-126). | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P21812 | |
Goat | |
Ammonium Sulfate Precipitation | |
RUO | |
17227 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction