Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCPyV large T-antigen Mouse anti-Human, MilliporeSigma™

Catalog No. MABF204425 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF2044-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF2044-25 Supplier MilliporeSigma Supplier No. MABF204425UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-MCPyV large T-antigen, clone CM2B4 is a mouse monoclonal antibody that detects MCPyV large T antigen and has been tested for use in Immunohistochemistry, Immunoprecipitation, and Western Blotting for the detection of Merkel cell polyomavirus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCPyV large T-antigen
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols N/A
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to RSRKPSSNASRGA sequence from the Merkel cell polyomavirus large T antigen exon 2 with a C-terminal cystiene.
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone CM2B4 is highly specific for MCPyV large T and 57kT isoforms, but does not detect MCPyV small T antigen.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.