Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCPyV T-antigen Mouse anti-Virus, Unconjugated, Clone: 2t2, MilliporeSigma™

Catalog No. MABF23161MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF23161MI 100 μL
MABF23162MI 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF23161MI Supplier MilliporeSigma Supplier No. MABF2316100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MCPyV T-antigen Monoclonal antibody specifically detects MCPyV T-antigen in Virus samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCPyV T-antigen
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2t2
Conjugate Unconjugated
Gene Alias Merkel cell polyomavirus T antigen;MCV Large T antigen
Host Species Mouse
Immunogen A linear peptide corresponding to 17 amino acids from the J-domain region present in both MCV LT and sT proteins.
Molecular Weight of Antigen ∼17 and 48 kDa observed; 28.16 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μL
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Test Specificity Clone 2t2 is a mouse monoclonal antibody that detects MCV LT and the 57 kDa T.
Target Species Virus
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.