Learn More
MCR Safety Crews™ Rachet Take-up Headgear and Faceshield
Economical protection provides comfort and convenience
$7.39 - $1262.00
Catalog Number | Mfr. No. | Dimensions (L x W) | Material | Product Type | No. per Case | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W) | Material | Product Type | No. per Case | Price | Quantity & Availability | ||
19-056456
|
MCR Safety
181560 |
20.3 x 39.3 cm (8 x 15.5 in.) | Polycarbonate | Faceshield | 75/Cs. |
Case of 75 for $868.00
|
|
||
19-022074
|
MCR Safety
181640 |
20.3 x 40.6 cm (8 x 16 in.) | PETG | Faceshield | 100/Cs. |
Case of 100 for $1,262.00
|
|
||
19-056447
|
MCR Safety
181540 |
20.3 x 39.3 cm (8 x 15.5 in.) | Polycarbonate | Faceshield | 100/Cs. |
Each for $7.39
Pack of 25 EA for $175.40
|
|
||
19-047491
|
MCR Safety
103 |
10.5 x 15.5 in. | Polycarbonate | Ratchet Headgear | 24/Cs. |
Each for $23.80
Case of 24 EA for $570.50
|
|
||
19-022054
|
MCR Safety
102 |
Aluminum | Faceshield Bracket for Hardhat | 24/Cs. | N/A | N/A | N/A | ||
Description
An assortment of brackets and faceshield sizes and materials make it a good bet that you can find an option just right for you.Compliance
Meets ANSI Z87.1+ standards.
Specifications
Clear |