Learn More
MCR Safety Crews™ Storm™ Safety Glasses
Catalog No. p-97246 Shop All MCR Safety Products
Click to view available options
Frame Color:
Black
Navy
Red
Lens Coating:
Scratch-resistant
Lens Tint:
Clear
Gray
This item is not returnable. View return policy
Catalog No. 19-003-044
Supplier: MCR Safety ST110
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Workers in the Storm™ can expect extreme comfort with this face-flattering protective eyewear from Crews™. Ratchet action temples combined with non-slip rubber head grips create a customized fit for virtually any head size.
  • Available in a variety of frame colors and lens combinations
  • Wraparound lens constructed of lightweight, scratch resistant polycarbonate, that filters 99.9% of U.V. rays, delivers advanced visual acuity and protection
  • Single wraparound dual focal point lens with integral side shields
  • 5 position ratchet action bayonet temples
  • Exclusive Duramass™ scratch resistant coating
  • Non-slip, soft temple material
Compliance
Spectacles meet ANSI Z87.1-1989 standards for optical clarity and impact resistance.
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance Spectacles meet ANSI Z87.1-1989 standards for optical clarity and impact resistance.
Lens Tint Clear
Frame Color Black
Lens Material Polycarbonate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.